Anti-FTbeta Rabbit Polyclonal Antibody [clone: E285]
BWRLBS3377
- Antibody type:Primary
- Antigen symbol:FT beta
- Clonality:Polyclonal
- Clone:E285
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 250-300 of Human FTbeta.
FT? (E285) pAb detects endogenous levels of FT? protein.
Type: Primary
Antigen: FTbeta
Clonality: Polyclonal
Clone: E285
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat