Anti-IL-1alpha Rabbit Polyclonal Antibody [clone: F8]
Catalog # BWRLBS2825
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IL-1 alpha
- Clonality:Polyclonal
- Clone:F8
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 91-140 of Human IL-1alpha.
IL-1? (F8) pAb detects endogenous levels of IL-1? protein
Type: Primary
Antigen: IL-1alpha
Clonality: Polyclonal
Clone: F8
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat