Anti-14-3-3 Theta Rabbit Polyclonal Antibody [clone: W228]
Catalog # BWRLBS1790
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 theta
- Clonality:Polyclonal
- Clone:W228
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 200-240 of Human 14-3-3 Theta.
14-3-3 ? (W228) pAb detects endogenous levels of 14-3-3 protein theta/tau.
Type: Primary
Antigen: 14-3-3 Theta
Clonality: Polyclonal
Clone: W228
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat