Anti-Ku-86 Rabbit Polyclonal Antibody [clone: S462]
Catalog # BWRLBS2692
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Ku-86
- Clonality:Polyclonal
- Clone:S462
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 450-480 of Human Ku-86.
Ku-86 (S462) pAb detects endogenous levels of Ku-86 protein.
Type: Primary
Antigen: Ku-86
Clonality: Polyclonal
Clone: S462
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human