Anti-GRB10 Rabbit Polyclonal Antibody [clone: L63]
Catalog # BWRLBS1748
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GRB10
- Clonality:Polyclonal
- Clone:L63
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 32-84 of Human GRB10.
GRB10 (L63) pAb detects endogenous levels of GRB10 protein.
Type: Primary
Antigen: GRB10
Clonality: Polyclonal
Clone: L63
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat