Anti-HLA-DOalpha Rabbit Polyclonal Antibody [clone: V111]
Catalog # BWRLBS3134
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HLA-DO alpha
- Clonality:Polyclonal
- Clone:V111
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 81-130 of Human HLA-DOalpha.
HLA-DO? (V111) pAb detects endogenous levels of HLA-DO? protein.
Type: Primary
Antigen: HLA-DOalpha
Clonality: Polyclonal
Clone: V111
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human