Anti-PDGFR-alpha Rabbit Polyclonal Antibody [clone: S1057]
Catalog # BWRLBS1684
Leverancier: Bioworld Technology
Specificaties
- Antibody type:Primary
- Antigen symbol:PDGFR-alpha
- Clonality:Polyclonal
- Clone:S1057
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specificaties
Over dit artikel
Synthetic peptide, corresponding to amino acids 1020-1070 of Human PDGFR-alpha.
PDGFR-? (S1057) pAb detects endogenous levels of PDGFR-? protein.
Type: Primary
Antigen: PDGFR-alpha
Clonality: Polyclonal
Clone: S1057
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat