Anti-Histone H1oo Rabbit Polyclonal Antibody [clone: P18]
Catalog # BWRLBS1656
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Histone H1oo
- Clonality:Polyclonal
- Clone:P18
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human Histone H1oo.
Histone H1oo (P18) pAb detects endogenous levels of Histone H1oo protein
Type: Primary
Antigen: Histone H1oo
Clonality: Polyclonal
Clone: P18
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human