Anti-Topo IIalpha Rabbit Polyclonal Antibody [clone: Q14]
BWRLBS3010
- Antibody type:Primary
- Antigen symbol:Topo II alpha
- Clonality:Polyclonal
- Clone:Q14
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminus of Human Topo IIalpha.
Topo II? (Q14) pAb detects endogenous levels of Topo II? protein.
Type: Primary
Antigen: Topo IIalpha
Clonality: Polyclonal
Clone: Q14
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human