Anti-ZBP-89 Rabbit Polyclonal Antibody [clone: E84]
Catalog # BWRLBS2510
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ZBP-89
- Clonality:Polyclonal
- Clone:E84
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 53-98 of Human ZBP-89.
ZBP-89 (E84) pAb detects endogenous levels of ZBP-89 protein.
Type: Primary
Antigen: ZBP-89
Clonality: Polyclonal
Clone: E84
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat