Anti-Dlx-4 Rabbit Polyclonal Antibody [clone: Q90]
BWRLBS2500
- Antibody type:Primary
- Antigen symbol:Dlx-4
- Clonality:Polyclonal
- Clone:Q90
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 84-104 of Human Dlx-4.
Dlx-4 (Q90) pAb detects endogenous levels of Dlx-4 protein.
Type: Primary
Antigen: Dlx-4
Clonality: Polyclonal
Clone: Q90
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat