Anti-VIP Rabbit Polyclonal Antibody [clone: V120]
Catalog # BWRLBS2919
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:VIP
- Clonality:Polyclonal
- Clone:V120
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 91-140 of Human VIP.
VIP (V120) pAb detects endogenous levels of VIP protein
Type: Primary
Antigen: VIP
Clonality: Polyclonal
Clone: V120
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human