Anti-Urotensin II Rabbit Polyclonal Antibody [clone: I97]
BWRLBS2918
- Antibody type:Primary
- Antigen symbol:Urotensin II
- Clonality:Polyclonal
- Clone:I97
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 71-120 of Human Urotensin II.
Urotensin II (I97) pAb detects endogenous levels of Urotensin II protein.
Type: Primary
Antigen: Urotensin II
Clonality: Polyclonal
Clone: I97
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human