Anti-Ku-86 Rabbit Polyclonal Antibody [clone: A717]
BWRLBS1497
- Antibody type:Primary
- Antigen symbol:Ku-86
- Clonality:Polyclonal
- Clone:A717
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 700-730 of Human Ku-86.
XRCC5 (A717) pAb detects endogenous levels of XRCC5 protein.
Type: Primary
Antigen: Ku-86
Clonality: Polyclonal
Clone: A717
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse