Anti-Nkx-3.1 Rabbit Polyclonal Antibody [clone: L2]
BWRLBS1456
- Antibody type:Primary
- Antigen symbol:Nkx-3.1
- Clonality:Polyclonal
- Clone:L2
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminual of Human Nkx-3.1.
NKX3.1 (L2) pAb detects endogenous levels of NKX3.1 protein.
Type: Primary
Antigen: Nkx-3.1
Clonality: Polyclonal
Clone: L2
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human