Anti-Cox-11 Rabbit Polyclonal Antibody [clone: F80]
Catalog # BWRLBS2185
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cox-11
- Clonality:Polyclonal
- Clone:F80
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 50-100 of Human Cox-11.
Cox-11 (F80) pAb detects endogenous levels of Cox-11 protein.
Type: Primary
Antigen: Cox-11
Clonality: Polyclonal
Clone: F80
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat