Anti-14-3-3 beta Rabbit Polyclonal Antibody [clone: Q69 ]
Catalog # BWRLBS2176
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 beta
- Clonality:Polyclonal
- Clone:Q69
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 40-90 of Human 14-3-3 beta.
14-3-3 ? (Q69) pAb detects endogenous levels of 14-3-3 ? protein.
Type: Primary
Antigen: 14-3-3 beta
Clonality: Polyclonal
Clone: Q69
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat