Order Entry
Netherlands
ContactUsLinkComponent
Anti-GAD-65/67 Rabbit Polyclonal Antibody [clone: D572]
Anti-GAD-65/67 Rabbit Polyclonal Antibody [clone: D572]
Catalog # BWRLBS1400
Anti-GAD-65/67 Rabbit Polyclonal Antibody [clone: D572]
Catalog # BWRLBS1400

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    GAD-65/67
  • Clonality:
    Polyclonal
  • Clone:
    D572
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 540-590 of Human GAD-65.

GAD-65/67 (D572) pAb detects endogenous levels of GAD65 and GAD67 protein.

Type: Primary
Antigen: GAD-65/67
Clonality: Polyclonal
Clone: D572
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat