Anti-Septin 1 Rabbit Polyclonal Antibody [clone: D205]
Catalog # BWRLBS2111
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Septin 1
- Clonality:Polyclonal
- Clone:D205
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 169-224 of Human Septin 1.
Septin 1 (D205) pAb detects endogenous levels of Septin 1 protein.
Type: Primary
Antigen: Septin 1
Clonality: Polyclonal
Clone: D205
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat