Anti-C/EBP-epsilon Rabbit Polyclonal Antibody [clone: K220]
Catalog # BWRLBS2074
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:C/EBP- epsilon
- Clonality:Polyclonal
- Clone:K220
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 188-245 of Human C/EBP-epsilon.
C/EBP-? (K220) pAb detects endogenous levels of C/EBP-? protein.
Type: Primary
Antigen: C/EBP-epsilon
Clonality: Polyclonal
Clone: K220
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat