Order Entry
Netherlands
ContactUsLinkComponent
Anti-ASC-1 Rabbit Polyclonal Antibody [clone: G105]
Anti-ASC-1 Rabbit Polyclonal Antibody [clone: G105]
Catalog # BWRLBS2072
Anti-ASC-1 Rabbit Polyclonal Antibody [clone: G105]
Catalog # BWRLBS2072

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ASC-1
  • Clonality:
    Polyclonal
  • Clone:
    G105
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 72-124 of Human ASC-1.

ASC-1 (G105) pAb detects endogenous levels of ASC-1 protein.

Type: Primary
Antigen: ASC-1
Clonality: Polyclonal
Clone: G105
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse