Anti-Dyrk1A Rabbit Polyclonal Antibody [clone: Q39]
Catalog # BWRLBS2054
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Dyrk1A
- Clonality:Polyclonal
- Clone:Q39
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 6-58 of Human Dyrk1A.
Dyrk1A (Q39) pAb detects endogenous levels of Dyrk1A protein.
Type: Primary
Antigen: Dyrk1A
Clonality: Polyclonal
Clone: Q39
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat