Anti-LT-beta Rabbit Polyclonal Antibody [clone: V211]
Catalog # BWRLBS2038
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:LT- beta
- Clonality:Polyclonal
- Clone:V211
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 200-250 of Human LT-beta.
LT-? (V211) pAb detects endogenous levels of LT-? protein.
Type: Primary
Antigen: LT-beta
Clonality: Polyclonal
Clone: V211
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse