Anti-SCAND1 Rabbit Polyclonal Antibody [clone: E88]
Catalog # BWRLBS2023
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:SCAND1
- Clonality:Polyclonal
- Clone:E88
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 50-100 of Human SCAND1.
SCAND1 (E88) pAb detects endogenous levels of SCAND1 rotein.
Type: Primary
Antigen: SCAND1
Clonality: Polyclonal
Clone: E88
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human