Order Entry
Netherlands
ContactUsLinkComponent
Anti-AF-10 Rabbit Polyclonal Antibody [clone: E237]
Anti-AF-10 Rabbit Polyclonal Antibody [clone: E237]
Catalog # BWRLBS2019
Anti-AF-10 Rabbit Polyclonal Antibody [clone: E237]
Catalog # BWRLBS2019

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    AF-10
  • Clonality:
    Polyclonal
  • Clone:
    E237
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 200-250 of Human AF-10.

AF-10 (E237) pAb detects endogenous levels of AF-10 protein.

Type: Primary
Antigen: AF-10
Clonality: Polyclonal
Clone: E237
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat