Anti-Cyclophilin F Rabbit Polyclonal Antibody [clone: Y121]
Catalog # BWRLBS1976
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cyclophilin F
- Clonality:Polyclonal
- Clone:Y121
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 91-140 of Human Cyclophilin F.
Cyclophilin F (Y121) pAb detects endogenous levels of Cyclophilin F protein.
Type: Primary
Antigen: Cyclophilin F
Clonality: Polyclonal
Clone: Y121
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat