Anti-GCS-beta-2 Rabbit Polyclonal Antibody [clone: V102]
BWRLBS3445
- Antibody type:Primary
- Antigen symbol:GCS- beta-2
- Clonality:Polyclonal
- Clone:V102
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 71-120 of Human GCS-beta-2.
GCS-?-2 (V102) pAb detects endogenous levels of GCS-?-2 protein.
Type: Primary
Antigen: GCS-beta-2
Clonality: Polyclonal
Clone: V102
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat