Anti-CYB561D2 Rabbit Polyclonal Antibody [clone: H86]
Catalog # BWRLBS3424
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CYB561D2
- Clonality:Polyclonal
- Clone:H86
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 61-110 of Human CYB561D2.
CYB561D2 (H86) pAb detects endogenous levels of CYB561D2 protein.
Type: Primary
Antigen: CYB561D2
Clonality: Polyclonal
Clone: H86
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat