Anti-Skp2 p45 Rabbit Polyclonal Antibody [clone: P399]
BWRLBS1325
- Antibody type:Primary
- Antigen symbol:Skp2 p45
- Clonality:Polyclonal
- Clone:P399
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 369-419 of Human Skp2 p45.
SKP2 p45 (P399) pAb detects endogenous levels of SKP2 p45 protein.
Type: Primary
Antigen: Skp2 p45
Clonality: Polyclonal
Clone: P399
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat