Order Entry
Netherlands
ContactUsLinkComponent
Anti-HnRNP F Rabbit Polyclonal Antibody
Anti-HnRNP F Rabbit Polyclonal Antibody
Catalog # BSBTA05806
Supplier:  Boster Bio
undefined
Anti-HnRNP F Rabbit Polyclonal Antibody
Catalog # BSBTA05806
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HnRNP F
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Heterogeneous nuclear ribonucleoprotein F(HNRNPF) detection. Tested with WB, IHC-P in Human, Mouse, Rat.

Type: Primary
Antigen: HnRNP F
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat