Ion source cleaning powder
Catalog # REST22685
Supplier: Restek
Specifications
- Description:Ion source cleaning powder, 1 kg
- Pk:1 kg
Specifications
About this item
This aluminum oxide powder is used to clean surfaces that contact the sample or ion beam when you encounter poor sensitivity and inadequate abundances at high masses.