Anti-Envelope Protein Mouse Monoclonal Antibody [clone: M86646]
Catalog # ABNOMAB14215
Supplier: Abnova
Specifications
- Type antilichaam:Primair
- antigeen symbool:Envelope Protein
- Clonality:Monoklonaal
- Clone:M86646
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Virus
- Formulier:Liquid
- Opslagbuffer:In 0.015 M potassium phosphate, 0.85% NaCl, pH 7.2 (0.05% sodium azide).
- Immunogeen:Recombinant protein corresponding to amino acids 291-690 of Zika virus envelope protein.
- Purification:DEAE column chromatography
- Maat:500 µG
- VPE:500 µG
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant Zika virus envelope protein.
Type: Primary
Antigen: Envelope Protein
Clonality: Monoclonal
Clone: M86646
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Viruses