Order Entry
Netherlands
ContactUsLinkComponent
Streptococcus sp. glycerol-3-phosphate oxidase, MP Biomedicals
Streptococcus sp. glycerol-3-phosphate oxidase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Streptococcus sp. glycerol-3-phosphate oxidase, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9046-28-0
  • Species:
    Streptococcus sp.
  • Storage conditions:
    −20 °C
  • Form:
    Yellow lyophilized powder
  • Enzyme name:
    Glycerol-3-Phosphate oxidase
  • Enzyme activity:
    >10 u/mg solid
  • Molecular weight:
    131 kDa
  • pH:
    7,5 - 8,0

Specifications

Frequently Bought Together