Capillary columns, Rxi®-17Sil MS columns (fused silica)
Leverancier: Restek
Certificaten
Over dit artikel
These fused silica columns have midpolarity Crossbond® phase manufactured for column to column reproducibility suited for validated methods.
- Inert column and highly selective for active environmental compounds such as PAHs
- Low bleed for use with sensitive detectors such as MS
- Equivalent to USP phase G3
Similar phases: DB-17ms, VF-17ms
Frequently Bought Together