Rat recombinant CNTF
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Species:Rat
- Storage conditions:Store at −20 °C.
- Biological activity:Determined by its ability to stimulate proliferation of human TF-1 cells using a concentration range of 25.0-35.0 ng/ml.
- Protein synonyms:Ciliary Neurotrophic Factor
- UniProtKB:P20294
- Protein/peptide name:CNTF
- Purity:>98%
- Solubility:It is recommended to reconstitute the lyophilized CNTF in sterile water or 0.4% NaHCO3 adjusted to pH 8-9, not less than 100ug/ml, which can then be further diluted to other aqueous solutions, preferably in presence of carrier protein.
- Sequence:AFAEQTPLTL HRRDLCSRSI WLARKIRSDL TALMESYVKH QGLNKNINLD SVDGVPVAST DRWSEMTEAE RLQENLQAYR TFQGMLTKLL EDQRVHFTPT EGDFHQAIHT LMLQVSAFAY QLEELMVLLE QKIPENEADG MPATVGDGGL FEKKLWGLKV LQELSQWTVR SIHDLRVISS HQMGISALES HYGAKDKQM
- Formulation:Lyophilized from concentrated (1mg/ml) solution in water containing 0.025% NaHCO3.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together