PDGF-AA
Catalogus # BIRBORB97821-10
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:10 µG
- Storage conditions:Store at−20 °C.
- Biological activity:ED50: Less than 1 ng/ml, corresponding to a specific activity of 1x 106 IU/mg, calculated by the dose dependent proliferation of murine BALB/c 3T3 indicator cells measured by3H-thymidine uptake.
- Protein synonyms:ODGF|GDGF
- UniProtKB:P04085
- Protein/peptide name:PDGF-AA
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized PDGF-AAin sterile bidest water not less than 100 g/ml. For longterm storage it is recommended to add a carrier protein(0.1% HSA or BSA).
- Molecular weight:29 kDa
- Formulation:Lyophilized