Order Entry
Netherlands
ContactUsLinkComponent
Human recombinant ENA 78 (from E. coli)
Human recombinant ENA 78 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant ENA 78 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The biological activity was determined by measuring the dose dependent mobilization of intracellular calcium (calcium flux) with human neutrophils. Significant calcium mobilization is observed with 100ng/mL of recombinant human ENA-78.
  • Protein synonyms:
    Small inducible cytokine B5
  • Protein/peptide name:
    ENA 78
  • Purity:
    > 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized ENA-78 in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be, Ala- Ala -Val-Leu-Arg.
  • Formulation:
    The CXCL5 was lyophilized from concentrated (1mg/ml) solution in water containing no additives.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together