Order Entry
Netherlands
ContactUsLinkComponent
Human EGF 21-Leu (from E. coli)
Human EGF 21-Leu (from E. coli)
Supplier:  Biorbyt
Certificates
Human EGF 21-Leu (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized Epidermal Growth Factor 21 Leu although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution EGF 21-Leu should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    The ED50, calculated by the dose-dependant proliferation of MDCK cells is 10 ng/ml.
  • Protein synonyms:
    Urogastrone
  • Protein/peptide name:
    EGF 21-Leu
  • Purity:
    Greater than 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Epidermal Growth Factor 21-Leu in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be Asn-Ser-Asp-Ser-Glu.
  • Formulation:
    The protein was lyophilized from concentrated (1mg/ml) solution with no additives.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together