Order Entry
Netherlands
ContactUsLinkComponent
Cylinder valve wrench
Cylinder valve wrench
Catalog # REST21321
Supplier:  Restek
undefined
Cylinder valve wrench
Catalog # REST21321
Supplier:  Restek
Specifications
  • Description:
    Wrench for cylinder valve
  • Pk:
    1
Specifications
About this item
This specially designed wrench enables easy opening of cylinder valves that are fitted with a hand wheel. It is also suitable for removing difficult cylinder caps.