Order Entry
Netherlands
ContactUsLinkComponent
Nuclear extract from HeLa cells
Nuclear extract from HeLa cells
Catalog # BMOLKI-140-0100
CAS Number:  
undefined
Nuclear extract from HeLa cells
Catalog # BMOLKI-140-0100
CAS Number:  
Specifications
  • Pk:
    100 µl
  • Description:
    Nuclear Extract from HeLa Cells, 100 µl
Specifications
About this item
Nuclear extract from HeLa cells is a rich source of HDAC activity which is used for inhibitor screening or immunoprecipitation of HDAC containing complexes.