Intermediate-polarity deactivated guard/retention gap columns/transfer lines (fused silica)
Supplier: Restek
Certificates
About this item
Intermediate-polarity deactivated guard/retention gap columns are useful for a wide range of applications and are proven to be compatible with most common solvents.
- Tested with a comprehensive test mix to ensure high inertness
- Useful for a wide range of applications
- Use with most common solvents
- Maximum temperature: 325 °C
- 30- and 60-meter lengths are banded in 5-meter sections
Frequently Bought Together