Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-PODXL Mouse Monoclonal Antibody [clone: 4F10]
  ABNOMAB14397
 :  Abnova
Anti-PODXL Mouse Monoclonal Antibody [clone: 4F10]
  ABNOMAB14397
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    PODXL Recombinant
  • Antigen symbol:
    PODXL
  • Clonality:
    Monoclonal
  • Clone:
    4F10
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    5420
  • Amino acid number:
    O00592
  • Storage buffer:
    In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
  • Immunogen:
    Recombinant protein corresponding to intracellular, transmembrane, and part of the extracellular domain of human PODXL.
  • Purification:
    PEG precipitation
  • Size:
    100 µG
  • Pk:
    100 µG

 

 

Mouse monoclonal antibody raised against partial recombinant human PODXL.

Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 4F10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human