Anti-MUC6 Mouse Monoclonal Antibody [clone: MUC6/916]
Catalog # ABNOMAB14369
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:MUC6 Recombinant
- Antigen symbol:MUC6
- Clonality:Monoclonal
- Clone:MUC6/916
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:4588
- Amino acid number:Q6W4X9
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Recombinant protein corresponding to full length human MUC6.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human MUC6.
Type: Primary
Antigen: MUC6
Clonality: Monoclonal
Clone: MUC6/916
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human