Order Entry
Netherlands
ContactUsLinkComponent
Anti-KRT7 Mouse Monoclonal Antibody [clone: OV-TL12/30]
Anti-KRT7 Mouse Monoclonal Antibody [clone: OV-TL12/30]
Catalog # ABNOMAB14308
Supplier:  Abnova
Anti-KRT7 Mouse Monoclonal Antibody [clone: OV-TL12/30]
Catalog # ABNOMAB14308
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 7
  • Antigen symbol:
    KRT7
  • Clonality:
    Monoclonal
  • Clone:
    OV-TL12/30
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    3855
  • Amino acid number:
    P08729
  • Storage buffer:
    In 10 mM PBS.
  • Immunogen:
    OTN 11 ovarian carcinoma cell line.
  • Purification:
    Protein A/G purification
  • Size:
    100 µG
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against native human KRT7.

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: OV-TL12/30
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human