Order Entry
Netherlands
ContactUsLinkComponent
Anti-SUMO1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: SUMO1/1188]
Anti-SUMO1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: SUMO1/1188]
  BTIUBNUM1188-50
 :  Biotium
Anti-SUMO1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: SUMO1/1188]
  BTIUBNUM1188-50
 :  Biotium

 

  • Antibody type:
    Primary
  • Antigen name:
    SUMO-1
  • Antigen symbol:
    Small ubiquitin-related modifier 1
  • Clonality:
    Monoclonal
  • Clone:
    Clone SUMO1/1188
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Gene ID:
    596171
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

 

 

This MAb is specific to SUMO-1 and shows no cross-reaction with either SUMO-2 or SUMO-3. The small ubiquitin-related modifier (SUMO) proteins, which include SUMO-1, SUMO-2 and SUMO-3, belong to the ubiquitin-like protein family. Like ubiquitin, the SUMO proteins are synthesized as precursor proteins that undergo processing before conjugation to target proteins. Also, both utilize the E1, E2, and E3 cascade enzymes for conjugation. However, SUMO and ubiquitin differ with respect to targeting. Ubiquitination predominantly targets proteins for degradation, whereas sumoylation targets proteins to a variety of cellular processing, including nuclear transport, transcriptional regulation, apoptosis and protein stability. The unconjugated SUMO-1 protein localizes to the nuclear membrane.

Type: Primary
Antigen: SUMO1
Clonality: Monoclonal
Clone: SUMO1/1188
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat