Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CDC20 Mouse Monoclonal Antibody (CF405S) [clone: CDC20/1102]
 :  Biotium
 
Anti-CDC20 Mouse Monoclonal Antibody (CF405S) [clone: CDC20/1102]
 :  Biotium

 

  • Catalog No:
  • BNC041102-500
  • BNC041102-100
  • Antigen symbol:
  • CDC20
  • CDC20
  • Antigen name:
  • Cell division cycle 20
  • Cell division cycle 20
  • Antigen synonyms:
  • P55CDC-LSB|CDC20A|p55CDC|CDC20
  • P55CDC-LSB|CDC20A|p55CDC|CDC20
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone CDC20/1102
  • Clone CDC20/1102
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 524947
  • 524947
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together