Order Entry
Netherlands
ContactUsLinkComponent
Anti-PTPRC Mouse Monoclonal Antibody (CF405S) [clone: DF-B1]
Anti-PTPRC Mouse Monoclonal Antibody (CF405S) [clone: DF-B1]
Supplier:  Biotium
Certificates
Anti-PTPRC Mouse Monoclonal Antibody (CF405S) [clone: DF-B1]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC041066-500
  • BNC041066-100
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • Antigen name:
  • Protein tyrosine phosphatase receptor type C
  • Protein tyrosine phosphatase receptor type C
  • Antigen synonyms:
  • Lyt-4|Leukocyte common antigen (LCA)|CD45R|B220|T200 glycoprotein|Ly-5|GP180|Protein tyrosine phosphatase receptor type C (PTPRC)|Receptor-type tyrosine-protein phosphatase C|Loc
  • Lyt-4|Leukocyte common antigen (LCA)|CD45R|B220|T200 glycoprotein|Ly-5|GP180|Protein tyrosine phosphatase receptor type C (PTPRC)|Receptor-type tyrosine-protein phosphatase C|Loc
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone DF-B1
  • Clone DF-B1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 654514
  • 654514
  • Isotype:
  • IgG3 kappa
  • IgG3 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together