Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD48 Mouse Monoclonal Antibody (Purified) [clone: 5-4.8]
Anti-CD48 Mouse Monoclonal Antibody (Purified) [clone: 5-4.8]
Supplier:  Biotium
Certificates
Anti-CD48 Mouse Monoclonal Antibody (Purified) [clone: 5-4.8]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB1057-100
  • BNUB1057-500
  • Antigen symbol:
  • CD48
  • CD48
  • Antigen synonyms:
  • BCM1 surface antigen|B cell membrane protein|SLAM family member 2 (SLAMF2)|BLAST1|Signaling lymphocytic activation molecule 2|TCT.1|B-cell activation marker
  • BCM1 surface antigen|B cell membrane protein|SLAM family member 2 (SLAMF2)|BLAST1|Signaling lymphocytic activation molecule 2|TCT.1|B-cell activation marker
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone 5-4.8
  • Clone 5-4.8
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 372679
  • 372679
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together