Order Entry
Netherlands
ContactUsLinkComponent
Anti-MS4A1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 93-1B3]
Anti-MS4A1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 93-1B3]
Catalog # BNUM1047-50
Supplier:  Biotium
Anti-MS4A1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 93-1B3]
Catalog # BNUM1047-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD20
  • Antigen symbol:
    MS4A1
  • Clonality:
    Monoclonal
  • Clone:
    Clone 93-1B3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Gene ID:
    712553
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Recognizes a protein of 30-33 kDa, which is identified as CD20 (Workshop V; Code CD20.4). It is a non-Ig differentiation antigen of B-cells and its expression is restricted to normal and neoplastic B-cells, being absent from all other leukocytes and tissues. CD20 is expressed by pre B-cells and persists during all stages of B-cell maturation but is lost upon terminal differentiation into plasma cells. The protein passes through the membrane 4 times with both ends in cytoplasm and exposes one short and one longer loop to the external environment. CD20 is not glycosylated in resting B-cells and its cytoplasmic domains are differentially phosphorylated upon activation. It acts as calcium channel involved in B cell activation and cell cycle progression.

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 93-1B3
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human