Order Entry
Netherlands
ContactUsLinkComponent
Anti-MS4A1 Mouse Monoclonal Antibody (Purified) [clone: 93-1B3]
Anti-MS4A1 Mouse Monoclonal Antibody (Purified) [clone: 93-1B3]
Leverancier:  Biotium
Certificaten
Anti-MS4A1 Mouse Monoclonal Antibody (Purified) [clone: 93-1B3]
Leverancier:  Biotium

Specificaties

  • Catalog No:
  • BNUB1047-100
  • BNUB1047-500
  • Antigen symbol:
  • MS4A1
  • MS4A1
  • Antigen name:
  • CD20
  • CD20
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone 93-1B3
  • Clone 93-1B3
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 186820
  • 186820
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specificaties

Frequently Bought Together